MilliporeSigma
New methylene homologation method for cyclic ketones.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-08-16)
Huaqing Liu, Chunrui Sun, Nam-Kyu Lee, Roger F Henry, Daesung Lee
ABSTRACT
Teaching new tricks to an old dog: By intercepting adducts between ketones and lithium trimethylsilyldiazomethane, a new Tiffeneau-Demjanov type methylene homologation could be realized in a single-step operation. Among proton sources and Lewis acids, silica gel was found to be the most effective reagent for the protonation of intermediates and their subsequent ring expansion (see scheme).