MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Highly efficient antiviral and antibacterial activities of solid-state cuprous compounds.
Highly efficient antiviral and antibacterial activities of solid-state cuprous compounds.
Journal of hazardous materials (2012-08-21)
Kayano Sunada, Masafumi Minoshima, Kazuhito Hashimoto
ABSTRACT
We found that several solid-state cuprous compounds, including cuprous oxide (Cu(2)O), sulfide (Cu(2)S), iodide (CuI), and chloride (CuCl), have highly efficient antiviral activities, whereas those of solid-state silver and cupric compounds are markedly lower. On a Cu(2)O-loaded glass substrate, for example, the infectious activity of bacteriophages was reduced by 5-orders of magnitude within 30 min and by 3-orders of magnitude within 1h for bacteria. In contrast, the infectious activities of both phages and bacteria were not markedly reduced on CuO-loaded substrates within a similar time frame. To determine the origin of this inhibitory activity, we investigated the effects of reactive oxygen species (ROS), leached copper ions, and the solid-state compound itself against bacteriophages, and concluded that infectious activity is lost following direct contact with the solid-state surface of cuprous compounds, but not ROS or copper ions. Furthermore, we found that Cu(2)O adsorbed and denatured more proteins than CuO, which suggests the difference of the inhibitory activity between Cu(2)O and CuO.
Sigma-Aldrich
Silver iodide, 99.999% trace metals basis
Sigma-Aldrich
Silver iodide, 99%
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis