MilliporeSigma
  • Preparation and characterization of novel oxidized cellulose acetate methyl esters.
Preparation and characterization of novel oxidized cellulose acetate methyl esters.
Carbohydrate polymers (2012-09-05)
D Yang, V Kumar
ABSTRACT
In this paper, we report the preparation of oxidized cellulose acetate methyl esters (OCAM) from OCA (OC14A: carboxylic acid content 10.6% (w/w), degree of acetyl group substitution: 1.89; OC21A: carboxylic acid content 15.7% (w/w), degree of acetyl group substitution: 1.70) by treatment with methanol at room temperature using 4-dimethylaminopyridine (DMAP) as a catalyst and dicyclohexylcarbodiimide (DCC) as a coupling agent. The new polymers were characterized by Fourier-transform infrared (FT-IR) and (1)H and (13)C nuclear magnetic resonance spectroscopies, carboxylic acid content determination, moisture sorption isotherms, intrinsic viscosity, and powder X-ray diffractometry. The new polymers are amorphous powders. It is practically insoluble in water but show solubility in a range of organic solvents.
Sigma-Aldrich
Cellulose acetate, average Mn ~30,000 by GPC
Sigma-Aldrich
Cellulose acetate, average Mn ~50,000 by GPC