MilliporeSigma
  • Beet juice-induced green fabrication of plasmonic AgCl/Ag nanoparticles.
Beet juice-induced green fabrication of plasmonic AgCl/Ag nanoparticles.
ChemSusChem (2012-09-05)
Jiahui Kou, Rajender S Varma
ABSTRACT
A simple, green, and fast approach (complete within 5 min) was explored for the fabrication of hybrid AgCl/Ag plasmonic nanoparticles under microwave (MW) irradiation. In this method, beet juice served as a reducing reagent, which is an abundant sugar-rich agricultural produce. Interestingly, the obtained AgCl/Ag samples have a smaller size than the AgCl reaction precursor. This makes it an unusual top-down hydrothermal synthesis. The as-prepared material displayed good photocatalytic activity for the degradation of methyl orange (MO) dye.
Sigma-Aldrich
Methyl Orange, suitable for microscopy
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Methyl Orange, JIS special grade
Sigma-Aldrich
Silver chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silver chloride, 99.999% trace metals basis
Sigma-Aldrich
Silver chloride, ReagentPlus®, 99%
Sigma-Aldrich
Methyl Orange, SAJ first grade
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4