MilliporeSigma
  • The cyclooxygenase and nitric oxide synthesis/pathways mediate the inhibitory serotonergic response to the pressor effect elicited by sympathetic stimulation in long-term diabetic pithed rats.
The cyclooxygenase and nitric oxide synthesis/pathways mediate the inhibitory serotonergic response to the pressor effect elicited by sympathetic stimulation in long-term diabetic pithed rats.
Pharmacology (2012-09-06)
B Restrepo, M García, C López, M L Martín, L San Román, A Morán
ABSTRACT
We investigated the mechanisms involved in the 5-hydroxytriptaminergic inhibitory action on the pressor responses elicited by sympathostimulation in long-term-diabetic pithed rats. Diabetes was induced in rats by alloxan administration. Eight weeks later, the animals were anaesthetized and pithed. The action and mechanisms of 5-HT were analysed based on the pressor responses induced by sympathostimulation. In 8-week-diabetic animals, 5-HT (20 µg/kg/min) inhibits the pressor effect of sympathostimulation which is reproduced by two selective 5-HT(1A) and 5-HT(2) receptor agonists: 8-hydroxydipropylaminotetralin hydrobromide (8-OH-DPAT, 5 µg/kg/min) and α-methyl-5-HT (5 µg/kg/min). A bolus injection of 1H-[1,2,4] oxadiazolo[4,3-a] quinoxalin-1-one (ODQ, 10 µg/kg), or L-arginine HCl, N(ω)-L-arginine methyl ester hydrochloride (L-NAME, 10 mg/kg), an inhibitor of NO production, prior to the infusion of 8-OH-DPAT (5 µg/kg/min) reversed the inhibitory effect of 8-OH-DPAT. The inhibitory effect of infusion of α-methyl 5-HT (5 µg/kg/min) was abolished in the presence of indomethacin (2 mg/kg), a non-selective cyclooxygenase (COX) inhibitor, or FR 122047 (1.5 mg/kg) or nimesulide (1.5 mg/kg), two selective COX-1 and COX-2 inhibitors, respectively, in long-term-diabetic pithed rats. Our results indicate that 5-HT inhibition of the pressor responses induced by electrical stimulation is mediated both by the NO and COX pathways in long-term-diabetic rats.
Sigma-Aldrich
(±)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide, ≥98%