MilliporeSigma
  • Bioaccumulation in freshwater crabs. Endosulfan accumulation in different tissues of Zilchiopsis collastinensis P. (Decapoda: Trichodactylidae).
Bioaccumulation in freshwater crabs. Endosulfan accumulation in different tissues of Zilchiopsis collastinensis P. (Decapoda: Trichodactylidae).
Bulletin of environmental contamination and toxicology (2012-09-11)
Carlos Leandro Negro, Lidia Eloísa Senkman, Jaquelina Vierling, María Rosa Repetti, Silvia Rosa García, Pablo Collins
ABSTRACT
We examined the bioaccumulation of α- and β-endosulfan and endosulfan sulfate in tissues from the crab Zilchiopsis collastinensis. There was more endosulfan accumulated in the hepatopancreas (from <2 to 467.8 ng g(-1)) than in the gonads (from <2 to 52.1 ng g(-1)) or muscles (<2 ng g(-1)). The endosulfan concentrations in the hepatopancreas decreased over time and with the endosulfan dilution (p < 0.05). In the gonads there was little bioaccumulation, which did not vary over time (p > 0.05). The hepatopancreas is a dynamic organ that is able to depurate itself, whereas the gonads act as a sink for pesticides.
Product Description
Supelco
Endosulfan, PESTANAL®, analytical standard