MilliporeSigma
  • Intestinal bacterium Eubacterium cellulosolvens deglycosylates flavonoid C- and O-glucosides.
Intestinal bacterium Eubacterium cellulosolvens deglycosylates flavonoid C- and O-glucosides.
Applied and environmental microbiology (2012-09-11)
Annett Braune, Michael Blaut
ABSTRACT
Eubacterium cellulosolvens cleaved the flavone C-glucosides homoorientin and isovitexin to their aglycones luteolin and apigenin, respectively. The corresponding isomers, orientin and vitexin, or other polyphenolic C-glucosides were not deglycosylated. E. cellulosolvens also cleaved several O-coupled glucosides of flavones and isoflavones to their corresponding aglycones.
Product Description
Isoorientin, primary reference standard
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)
Isovitexin, primary reference standard
Sigma-Aldrich
Isovitexin, ≥98.0% (HPLC)