MilliporeSigma
  • Farnesol induces apoptosis-like phenotype in the phytopathogenic fungus Botrytis cinerea.
Farnesol induces apoptosis-like phenotype in the phytopathogenic fungus Botrytis cinerea.
Mycologia (2012-09-11)
Milena Cotoras, Paulo Castro, Herman Vivanco, Ricardo Melo, Leonora Mendoza
ABSTRACT
This study demonstrates that the isoprenoid farnesol produces a toxic effect on the phytopathogenic fungus Botrytis cinerea in solid and liquid media. In solid media farnesol retarded 72 h the beginning of mycelial growth. Also, it was demonstrated that the toxic effect is due to farnesol triggers apoptosis in B. cinerea because ROS accumulation, DNA fragmentation and phosphatidylserine externalization were detected in farnesol-treated mycelium. Therefore, compounds that increase the intracellular farnesol or induce apoptosis could have a potential application as fungicide against B. cinerea.
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%