MilliporeSigma
  • Optimization of ionic liquid based ultrasonic assisted extraction of puerarin from Radix Puerariae Lobatae by response surface methodology.
Optimization of ionic liquid based ultrasonic assisted extraction of puerarin from Radix Puerariae Lobatae by response surface methodology.
Food chemistry (2012-09-18)
Jie-Ping Fan, Jing Cao, Xue-Hong Zhang, Jun-Zhong Huang, Tao Kong, Sheng Tong, Zhe-You Tian, Yan-Long Xie, Rui Xu, Jian-Hang Zhu
ABSTRACT
Ionic liquid (IL) based ultrasonic assisted extraction (ILUAE) was developed for the effective extraction of puerarin from Radix Puerariae Lobatae (RPL). The ILUAE parameters including the type of ILs, IL concentration, RPL amount, ultrasonic power and time were optimized by single-factor experiment and response surface methodology. Under the optimized experimental conditions, the best results were obtained using RPL amount 0.43 g in 10 mL 1.06 molL(-1) 1-butyl-3-methylimidazolium bromide aqueous solution, ultrasonic time 27.43 min and ultrasonic power 480 W. Scanning electron microscope images of RPL samples were obtained to provide visual evidence of the sonication effect. Compared with the conventional ultrasonic assisted extraction and refluent extraction, the proposed ILUAE offered shorter extraction time and remarkable higher efficiencies due to the higher penetration ability and solubility of IL and the cavitation phenomenon produced in the solvent by the passage of an ultrasonic wave, which further supported the suitability of the proposed approach.
Product Description
Supelco
Puerarin, analytical standard
Sigma-Aldrich
Puerarin, ≥98.0% (HPLC)