MilliporeSigma
  • Highly selective Barbier-type propargylations and allenylations catalyzed by titanocene(III).
Highly selective Barbier-type propargylations and allenylations catalyzed by titanocene(III).
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-09-22)
Juan Muñoz-Bascón, Iris Sancho-Sanz, Enrique Álvarez-Manzaneda, Antonio Rosales, J Enrique Oltra
ABSTRACT
The alkyne functional group is found in many bioactive natural products and is the key to many important chemical transformations developed over recent years. Moreover, allenes have recently gained relevance as versatile reagents in organic synthesis. Mild, catalytic methods to enable the selective introduction of either alkyne or allene motifs into organic molecules are very valuable but, as yet, quite scarce. We describe an extremely mild and selective method for either the propargylation or allenylation of carbonyl compounds catalyzed by the abundant, safe, and inexpensive metal titanium. These reactions can selectively provide homopropargylic alcohols from aldehydes and ketones or α-hydroxy-allenes from aldehydes. The mechanisms involved were also investigated.
Sigma-Aldrich
Allene, ≥95%