MilliporeSigma
  • Genome sequences of Pseudomonas luteola XLDN4-9 and Pseudomonas stutzeri XLDN-R, two efficient carbazole-degrading strains.
Genome sequences of Pseudomonas luteola XLDN4-9 and Pseudomonas stutzeri XLDN-R, two efficient carbazole-degrading strains.
Journal of bacteriology (2012-09-27)
Xiaorui Liu, Zhonghui Gai, Fei Tao, Hao Yu, Hongzhi Tang, Ping Xu
ABSTRACT
Pseudomonas luteola XLDN4-9 and Pseudomonas stutzeri XLDN-R are two efficient carbazole-degrading pseudomonad strains. Here we present 4.63- and 4.70-Mb assemblies of their genomes. Their annotated key genes for carbazole catabolism are similar, which may provide further insights into the molecular mechanism of carbazole degradation in Pseudomonas.
Product Description
Supelco
Carbazole, VETRANAL®, analytical standard
Sigma-Aldrich
Carbazole, ≥95% (GC)