MilliporeSigma
  • New, simple medium for selective recovery of Klebsiella pneumoniae and Klebsiella oxytoca from human feces.
New, simple medium for selective recovery of Klebsiella pneumoniae and Klebsiella oxytoca from human feces.
Journal of clinical microbiology (1984-11-01)
E Van Kregten, N A Westerdaal, J M Willers
ABSTRACT
A culture medium was developed which selectively favored the growth of Klebsiella pneumoniae and Klebsiella oxytoca in Escherichia coli-rich fecal cultures, without the use of antibiotics. The discriminative capacity of this medium was based on the presence of only two carbon sources, citrate and inositol, which can be utilized by nearly all K. pneumoniae and K. oxytoca strains but not by E. coli. The medium consisted of Simmons citrate agar (SCA) with 1% inositol (SCAI). Klebsiella strains from fecal samples subcultured on SCAI grew unhampered as yellow, dome-shaped, often mucoid colonies, whereas E. coli appeared as tiny, watery colonies. Apart from some Enterobacter strains, no other types of bacteria were found to mimic the typical appearance of klebsiellae. Recovery experiments from stool samples revealed a limiting ratio of Klebsiella to E. coli of 1:10(6) or more when samples were plated on SCAI versus ratios of 1:10(2) to 1:10(3) on blood agar or Macconkey agar. Compared with an existing Klebsiella culture method, the combination of SCA and MacConkey-inositol-carbenicillin (MIC) agar, Klebsiella yields with SCAI were not lower than those with the combination of MIC and SCA. Furthermore, the efficiency of the SCAI method was twice that of the latter combination. The SCAI plate could be a valuable tool in studies on the epidemiology of K. pneumoniae and K. oxytoca, for example in nosocomial infections, especially those concerning immunocompromised patients.
Product Description
Millipore
Simmons Citrate Agar, NutriSelect® Basic, powder, for coliforms, for yeasts, for molds, pack of 500 g, suitable for microbiology