MilliporeSigma
  • Novel spectrophotometric method for determination of some macrolide antibiotics in pharmaceutical formulations using 1,2-naphthoquinone-4-sulphonate.
Novel spectrophotometric method for determination of some macrolide antibiotics in pharmaceutical formulations using 1,2-naphthoquinone-4-sulphonate.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-10-09)
Safwan Ashour, Roula Bayram
ABSTRACT
New, simple and rapid spectrophotometric method has been developed and validated for the assay of two macrolide drugs, azithromycin (AZT) and erythromycin (ERY) in pure and pharmaceutical formulations. The proposed method was based on the reaction of AZT and ERY with sodium 1,2-naphthoquinone-4-sulphonate (NQS) in alkaline medium at 25 °C to form an orange-colored product of maximum absorption peak at 452 nm. All variables were studied to optimize the reaction conditions and the reaction mechanism was postulated. Beer's law was obeyed in the concentration range 1.5-33.0 and 0.92-8.0 μg mL(-1) with limit of detection values of 0.026 and 0.063 μg mL(-1) for AZT and ERY, respectively. The calculated molar absorptivity values are 4.3 × 10(4) and 12.3 × 10(4) L mol(-1) cm(-1) for AZT and ERY, respectively. The proposed methods were successfully applied to the determination of AZT and ERY in formulations and the results tallied well with the label claim. The results were statistically compared with those of an official method by applying the Student's t-test and F-test. No interference was observed from the concomitant substances normally added to preparations.