MilliporeSigma
  • Production of large graphene sheets by exfoliation of graphite under high power ultrasound in the presence of tiopronin.
Production of large graphene sheets by exfoliation of graphite under high power ultrasound in the presence of tiopronin.
Chemical communications (Cambridge, England) (2012-10-24)
Mildred Quintana, Marek Grzelczak, Konstantinos Spyrou, Bart Kooi, Sara Bals, Gustaaf Van Tendeloo, Petra Rudolf, Maurizio Prato
ABSTRACT
Under ultrasonication, the production of high quality graphene layers by exfoliation of graphite was achieved via addition of tiopronin as an antioxidant.
Sigma-Aldrich
N-(2-Mercaptopropionyl)glycine