- Development of Zn-ProPhenol-catalyzed asymmetric alkyne addition: synthesis of chiral propargylic alcohols.
Development of Zn-ProPhenol-catalyzed asymmetric alkyne addition: synthesis of chiral propargylic alcohols.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-10-26)
Barry M Trost, Mark J Bartlett, Andrew H Weiss, Axel Jacobi von Wangelin, Vincent S Chan
PMID23097281
ABSTRACT
The development of a general and practical zinc-catalyzed enantioselective alkyne addition methodology is reported. The commercially available ProPhenol ligand (1) has facilitated the addition of a wide range of zinc alkynylides to aryl, aliphatic, and α,β-unsaturated aldehydes in high yield and enantioselectivity. New insights into the mechanism of this reaction have resulted in a significant reduction in reagent stoichiometry, enabling the use of precious alkynes and avoiding the use of excess dimethylzinc. The enantioenriched propargylic alcohols from this reaction serve as versatile synthetic intermediates and have enabled efficient syntheses of several complex natural products.
MATERIALS