MilliporeSigma
  • Capillary electrophoresis to quantitate gossypol enantiomers in cotton flower petals and seed.
Capillary electrophoresis to quantitate gossypol enantiomers in cotton flower petals and seed.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-11-06)
Sergey Vshivkov, Egor Pshenichnov, Zamira Golubenko, Alik Akhunov, Shadman Namazov, Robert D Stipanovic
ABSTRACT
Gossypol is a toxic compound that occurs as a mixture of enantiomers in cotton plant tissues including seed and flower petals. The (-)-enantiomer is more toxic to non-ruminant animals. Efforts to breed cottonseed with a low percentage of (-)-gossypol requires determination of the (+)- to (-)-gossypol ratio in seed and flower petals. We report a method to quantitatively determine the total gossypol and percent of its enantiomers in cotton tissues using high performance capillary electrophoresis (HPCE). The method utilizes a borate buffer at pH 9.3 using a capillary with internal diameter of 50μm, effective length of 24.5cm, 15kV and cassette temperature of 15°C. This method provides high accuracy and reproducible results with a limit of detection of the individual enantiomers of less than 36ng/mL providing base line separation in less than 6min.
Sigma-Aldrich
(±)-Gossypol from cotton seeds, ≥95% (HPLC)