MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Oxidative difunctionalization of 2-amino-4H-pyrans in iodobenzene diacetate and N-chlorosuccinimide: reactivity, mechanistic insights, and DFT calculations.
Oxidative difunctionalization of 2-amino-4H-pyrans in iodobenzene diacetate and N-chlorosuccinimide: reactivity, mechanistic insights, and DFT calculations.
The Journal of organic chemistry (2012-11-08)
Santhosh Reddy Mandha, Manjula Alla, Vittal Rao Bommena, Jagadeesh Babu Nanubolu, Santosh Kumar Lingala, Soujanya Yarasi
ABSTRACT
Oxidative difunctionalization of 2-amino-4H-pyrans was accomplished with iodobenzene diacetate (IBD) and N-chlorosuccinimide (NCS) reagents in alcoholic medium. 2-Amino-4H-pyrans undergo geminal dialkoxylation with the migration of an amino group (1a,b, 2a-i, 3a,b, and 4) in IBD, whereas with NCS addition of both chlorine and alkoxy groups takes place across the chromene double bond (6a-i).
Sigma-Aldrich
Iodobenzene, 98%
Sigma-Aldrich
N-Chlorosuccinimide, 98%