MilliporeSigma
  • Fitness evaluation of two Brazilian Aedes aegypti field populations with distinct levels of resistance to the organophosphate temephos.
Fitness evaluation of two Brazilian Aedes aegypti field populations with distinct levels of resistance to the organophosphate temephos.
Memorias do Instituto Oswaldo Cruz (2012-11-14)
Thiago Affonso Belinato, Ademir Jesus Martins, Denise Valle
ABSTRACT
In Brazil, decades of dengue vector control using organophosphates and pyrethroids have led to dissemination of resistance. Although these insecticides have been employed for decades against Aedes aegypti in the country, knowledge of the impact of temephos resistance on vector viability is limited. We evaluated several fitness parameters in two Brazilian Ae. aegypti populations, both classified as deltamethrin resistant but with distinct resistant ratios (RR) for temephos. The insecticide-susceptible Rockefeller strain was used as an experimental control. The population presenting the higher temephos resistance level, Aparecida de Goiânia, state of Goiás (RR(95) of 19.2), exhibited deficiency in the following four parameters: blood meal acceptance, amount of ingested blood, number of eggs and frequency of inseminated females. Mosquitoes from Boa Vista, state of Roraima, the population with lower temephos resistance level (RR(95) of 7.4), presented impairment in only two parameters, blood meal acceptance and frequency of inseminated females. These results indicate that the overall fitness handicap was proportional to temephos resistance levels. However, it is unlikely that these disabilities can be attributed solely to temephos resistance, since both populations are also resistant to deltamethrin and harbour the kdr allele, which indicates resistance to pyrethroids. The effects of reduced fitness in resistant populations are discussed.
Product Description
Supelco
Temephos, PESTANAL®, analytical standard