MilliporeSigma
  • Hydroxysafflor yellow a inhibits lipopolysaccharide-induced inflammatory signal transduction in human alveolar epithelial A549 cells.
Hydroxysafflor yellow a inhibits lipopolysaccharide-induced inflammatory signal transduction in human alveolar epithelial A549 cells.
Fitoterapia (2012-11-20)
Lijuan Song, Yu Zhu, Ming Jin, Baoxia Zang
ABSTRACT
Hydroxysafflor yellow A (HSYA) is an active ingredient obtained from the flower of Carthamus tinctorius L. The present study investigated the effects of HSYA on lipopolysaccharide (LPS)-induced inflammatory signal transduction in human alveolar epithelial A549 cells. A549 cells stimulated with LPS were incubated with three doses of HSYA (1, 4 and 16μmol/L). HSYA suppressed the expression of TLR-4, Myd88, ICAM-1, TNFα, IL-1β and IL-6 at the mRNA and protein level, and inhibited the adhesion of leukocytes to A549 cells. HSYA treatment also decreased NF-κB p65 nuclear translocation and inhibited the phosphorylation of p38 mitogen-activated protein kinase (p38 MAPK). These findings suggest that HSYA effectively inhibits LPS-induced inflammatory signal transduction in A549 cells.