MilliporeSigma
  • A series of anti-CEA/anti-DOTA bispecific antibody formats evaluated for pre-targeting: comparison of tumor uptake and blood clearance.
A series of anti-CEA/anti-DOTA bispecific antibody formats evaluated for pre-targeting: comparison of tumor uptake and blood clearance.
Protein engineering, design & selection : PEDS (2012-11-24)
Paul J Yazaki, Brian Lee, Divya Channappa, Chia-Wei Cheung, Desiree Crow, Junie Chea, Erasmus Poku, Lin Li, Jan Terje Andersen, Inger Sandlie, Kelly Davis Orcutt, K Dane Wittrup, John E Shively, Andrew Raubitschek, David Colcher
ABSTRACT
A series of anti-tumor/anti-chelate bispecific antibody formats were developed for pre-targeted radioimmunotherapy. Based on the anti-carcinoembryonic antigen humanized hT84.66-M5A monoclonal antibody and the anti-DOTA C8.2.5 scFv antibody fragment, this cognate series of bispecific antibodies were radioiodinated to determine their tumor targeting, biodistribution and pharmacokinetic properties in a mouse xenograft tumor model. The in vivo biodistribution studies showed that all the bispecific antibodies exhibited specific high tumor uptake but the tumor targeting was approximately one-half of the parental anti-CEA mAb due to faster blood clearance. Serum stability and FcRn studies showed no apparent reason for the faster blood clearance. A dual radiolabel biodistribution study revealed that the (111)In-DOTA bispecific antibody had increased liver and spleen uptake, not seen for the (125)I-version due to metabolism and release of the radioiodine from the cells. These data suggest increased clearance of the antibody fusion formats by the mononuclear phagocyte system. Importantly, a pre-targeted study showed specific tumor uptake of (177)Lu-DOTA and a tumor : blood ratio of 199 : 1. This pre-targeted radiotherapeutic and substantial reduction in the radioactive exposure to the bone marrow should enhance the therapeutic potential of RIT.
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)