MilliporeSigma
  • Ultrasound assisted enzyme catalyzed transesterification of waste cooking oil with dimethyl carbonate.
Ultrasound assisted enzyme catalyzed transesterification of waste cooking oil with dimethyl carbonate.
Ultrasonics sonochemistry (2012-11-28)
Nikhil Gharat, Virendra K Rathod
ABSTRACT
This work reports the production of biodiesel with waste cooking oil and dimethyl carbonate in solvent free system through transesterification by immobilized enzyme (Novozym 435) under the influence of ultrasound irradiation. The experiments were conducted in an ultrasonic water bath under three different conditions i.e. ultrasonic irradiation (UI) without stirring, UI coupled with stirring and only stirring to compare their overall effects on fatty acid methyl esters (FAME) conversion. As compared with the conventional stirring method, where FAME conversion was 38.69% at 4h, the UI without stirring significantly enhanced the conversion of enzymatic transesterification to 57.68% for the same reaction time. However the reaction rate was further increased under the condition of ultrasonication coupled with stirring and resulted into higher conversion of 86.61% for the same reaction time. Effects of reaction parameters, such as temperature, ratio of DMC/oil, speed of agitation and enzyme loading on the conversion were investigated. Furthermore, repeated use of Novozym 435 showed gradual decline in both conversion as well as enzyme activity.
Sigma-Aldrich
Dimethyl carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl carbonate, anhydrous, ≥99%