MilliporeSigma
  • Dose intensification of busulfan in the preparative regimen is associated with improved survival: a phase I/II controlled, randomized study.
Dose intensification of busulfan in the preparative regimen is associated with improved survival: a phase I/II controlled, randomized study.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation (2012-12-12)
Simrit Parmar, Gabriela Rondon, Marcos de Lima, Peter Thall, Ronald Bassett, Paolo Anderlini, Partow Kebriaei, Issa Khouri, Prasanth Ganesan, Richard Champlin, Sergio Giralt
ABSTRACT
Dose intensity is important for disease control in patients undergoing allogeneic stem cell transplantation. We conducted a phase I/II controlled, adoptive, randomized study to determine the optimal dosing schedule of i.v. busulfan. Patients aged ≤75 years with advanced hematologic malignancies with human leukocyte antigen-compatible donor were eligible. All patients received fludarabine at 30 mg/m(2)/d for 4 days, and busulfan was administered in different doses in oral or i.v. formulations. As determined by the phase I trial, i.v. busulfan at a dose of 11.2 mg/kg/d was used for the phase II expansion cohort. Altogether, 80 patients with a median age of 56 years were enrolled. Forty percent had active disease at the time of transplantation. Engraftment occurred in 91%, and a complete response was achieved in 79% of patients posttransplantation. At a median follow-up of 91 months in the surviving patients, the outcomes for i.v. busulfan dose of 11.2 mg/kg/d versus other doses were as follows: nonrelapse mortality, 34% versus 23% (P = .4); cumulative incidence of relapse, 43% versus 68% (P = .02); relapse-free survival, 25% versus 9% (P = .017); and overall survival, 27% versus 9% (P = .02). We conclude that optimizing i.v. busulfan dose intensity in the preparative regimen may overcome disease-associated poor prognostic factors.
Product Description
Supelco
Busulfan, analytical standard, for drug analysis
Busulfan, European Pharmacopoeia (EP) Reference Standard