MilliporeSigma
  • A combined spectroscopic, docking and molecular dynamics simulation approach to probing binding of a Schiff base complex to human serum albumin.
A combined spectroscopic, docking and molecular dynamics simulation approach to probing binding of a Schiff base complex to human serum albumin.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-12)
N Fani, A K Bordbar, Y Ghayeb
ABSTRACT
The molecular mechanism of a Schiff base complex ((E)-((E)-2-(3-((E)-((E)-3(mercapto (methylthio) methylene)cyclopentylidene) amino) propylimino) cyclopentylidene) (methylthio) methanethiol) binding to Human Serum Albumin (HSA) was investigated by fluorescence quenching, absorption spectroscopy, molecular docking and molecular dynamics (MD) simulation procedures. The fluorescence emission of HSA was quenched by this Schiff base complex that has been analyzed for estimation of binding parameters. The titration of Schiff base solution by various amount of HSA was also followed by UV-Vis absorption spectroscopy and the corresponding data were analyzed by suitable models. The results revealed that this Schiff base has an ability to bind strongly to HSA and formed 1:1 complex. Energy transfer mechanism of quenching was discussed and the value of 5.45 ± 0.06 nm was calculated as the mean distance between the bound complex and the Trp residue. This is implying the high possibility of energy transfer from HSA to this Schiff base complex. Molecular docking results indicated that the main active binding site for this Schiff base complex is site III in subdomain IB. Moreover, MD simulation results suggested that this Schiff base complex can interact with HSA, without affecting the secondary structure of HSA but probably with a slight modification of its tertiary structure. MD simulations, molecular docking and experimental data reciprocally supported each other.
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)
Sigma-Aldrich
Sodium thiomethoxide, 95%