MilliporeSigma
Dichlorobenzoquinone chloroimine colorimetry of uric acid in urine.
Clinical chemistry (1990-04-01)
P Miller, V Oberholzer
ABSTRACT
In this nonenzymatic colorimetric method, dichlorobenzoquinone chloroimine is used as a color reagent to measure uric acid in urine. This method is less subject to the interferences that make enzymatic methods unreliable and is inexpensive, simple, and fast.
Product Description
Supelco
2,6-Dichloroquinone-4-chloroimide, for spectrophotometric det. of vitamin B6, ≥99.0%
Sigma-Aldrich
2,6-Dichloroquinone-4-chloroimide, ≥95% (TLC), suitable for detection of phenols