MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis of cyclopentanyl carbocyclic 5-fluorocytosine ((-)-5-fluorocarbodine) using a facially selective hydrogenation approach.
Synthesis of cyclopentanyl carbocyclic 5-fluorocytosine ((-)-5-fluorocarbodine) using a facially selective hydrogenation approach.
The Journal of organic chemistry (2012-12-13)
Jong Hyun Cho, Franck Amblard, Steven J Coats, Raymond F Schinazi
ABSTRACT
An efficient synthetic route to biologically relevant (-)-5-fluorocarbodine 6 was developed. Direct coupling of N(6)-protected 5-fluorouracil 15 with cyclopentenyl intermediate 13, followed by formation of a macrocycle between the base and the carbocyclic sugar moiety, via ring-closing metathesis, allowed for a facial selective hydrogenation of the sugar double bond to give, exclusively, the desired 4'-β stereoisomer.
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Cytidine, 99%