MilliporeSigma
Protein prosthesis: β-peptides as reverse-turn surrogates.
Protein science : a publication of the Protein Society (2012-12-15)
Ulrich Arnold, Bayard R Huck, Samuel H Gellman, Ronald T Raines
ABSTRACT
The introduction of non-natural modules could provide unprecedented control over folding/unfolding behavior, conformational stability, and biological function of proteins. Success requires the interrogation of candidate modules in natural contexts. Here, expressed protein ligation is used to replace a reverse turn in bovine pancreatic ribonuclease (RNase A) with a synthetic β-dipeptide: β²-homoalanine-β³-homoalanine. This segment is known to adopt an unnatural reverse-turn conformation that contains a 10-membered ring hydrogen bond, but one with a donor-acceptor pattern opposite to that in the 10-membered rings of natural reverse turns. The RNase A variant has intact enzymatic activity, but unfolds more quickly and has diminished conformational stability relative to native RNase A. These data indicate that hydrogen-bonding pattern merits careful consideration in the selection of beneficial reverse-turn surrogates.
Sigma-Aldrich
(R)-(–)-3-Piperidinecarboxylic acid, 97%
Sigma-Aldrich
D-2-Aminobutyric acid, 98%
Sigma-Aldrich
(S)-(+)-3-Piperidinecarboxylic acid, 97%
Sigma-Aldrich
L-2-Aminobutyric acid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Sigma-Aldrich
DL-2-Aminobutyric acid, ReagentPlus®, 99%