MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Studies on the inclusion behavior of 9-Aminoacridine into cyclodextrins: spectroscopic and theoretical evidences.
Studies on the inclusion behavior of 9-Aminoacridine into cyclodextrins: spectroscopic and theoretical evidences.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-22)
C Manivannan, R Vijay Solomon, P Venuvanalingam, R Renganathan
ABSTRACT
9-Aminoacridine (9-AA) is an important attractive pharmaceutical drug employed as chemotheraptic agent for wound dressings. However, 9-AA possesses limited solubility and rapid metabolic decomposition renders this potential drug to limit its applications. Here we propose cyclodextrins (CDs) as a drug carrier to improve the bioavailability, solubility of 9-AA. The interaction between 9-AA and CDs (α-CD and β-CD) has been studied using UV-Vis absorption, steady state time resolved fluorescence, (1)H NMR and FT-IR spectroscopy techniques. The spectroscopic measurements show that 9-AA does not form stable complex with α-CD and also confirmed by DFT calculations. On the other hand, 9-AA forms inclusion complex with β-CD in a 1:1 stoichiometry ratio. Our DFT results suggest that 9-AA stabilizes inside the CD environment through hydrogen bonding that has unambiguously confirmed by AIM analysis. Thus our studies provide a useful insights in the development of Aminoacridine based drugs & its delivery through a suitable carrier like CDs.
Product Description
Supelco
9-Aminoacridine, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)