MilliporeSigma
  • Identification of a sphingosine-sensitive Ca2+ channel in the plasma membrane of Leishmania mexicana.
Identification of a sphingosine-sensitive Ca2+ channel in the plasma membrane of Leishmania mexicana.
Biochemical and biophysical research communications (2012-12-25)
Gustavo Benaim, Yael García-Marchán, Claudia Reyes, Graciela Uzcanga, Katherine Figarella
ABSTRACT
The disruption of the intracellular Ca(2+) homeostasis of Leishmania mexicana represents a major target for the action of drugs, such as amiodarone and miltefosine. However, little is known about the mechanism of Ca(2+) entry to these cells. Here we show the presence of a Ca(2+) channel in the plasma membrane of these parasites. This channel has many characteristics similar to the human L-type voltage-gated Ca(2+) channel. Thus, Ca(2+) entry is blocked by verapamil, nifedipine and diltiazem while Bay K 8644 opened this channel. However, different to its human counterpart, sphingosine was able to open this channel, while other well known sphingolipids had no effect. This fact could have important pharmacological implications.
Sigma-Aldrich
(S)-(−)-Bay K8644, ≥98% (HPLC), solid