MilliporeSigma
  • The hybrid between the ABC domains of synapsin and the B subunit of Escherichia coli heat-labile toxin ameliorates experimental autoimmune encephalomyelitis.
The hybrid between the ABC domains of synapsin and the B subunit of Escherichia coli heat-labile toxin ameliorates experimental autoimmune encephalomyelitis.
Cellular immunology (2012-12-25)
Mario J Bibolini, M Julia Scerbo, Nahuel Peinetti, German A Roth, Clara G Monferran
ABSTRACT
The B subunit of Escherichia coli heat-labile enterotoxin (LTB) acts as efficient mucosal carrier for conjugated antigens. We expressed two heterologous proteins using E. coli as a host: a hybrid consisting of LTB and the A, B and C domain of synapsin (LTBABC) and the separated ABC peptide of this synaptic protein. Refolded LTBABC and LTB bound to the GM1 receptor and internalized into CHO-K1(GM1+) cells. LTBABC showed enhanced solubility and cell binding ability respect to the former hybrid LTBSC. Several oral doses of LTBABC were administered to rats with experimental autoimmune encephalomyelitis (EAE) from induction to the acute stage of the disease. This treatment decreased disease severity, delayed type hypersensitivity reaction and lymph node cell proliferation stimulated by myelin basic protein. Amelioration of EAE was also associated with modulation of the Th1/Th2 cytokine ratio, increased TGF-β secretion in mesenteric lymph nodes as well as expansion of CD4(+)CD25(+)Foxp3(+) regulatory T cell population. These results indicate that the fusion protein LTBABC is suitable for further exploration of its therapeutic effect on EAE development.
Sigma-Aldrich
Monosialoganglioside GM1 from bovine brain, lyophilized powder, BioXtra, γ-irradiated, ≥95% (TLC)
Sigma-Aldrich
Monosialoganglioside GM1 from bovine brain, ≥95%, lyophilized powder