MilliporeSigma
  • Enhancement of adhesive strength of hydroxyapatite films on Ti-29Nb-13Ta-4.6Zr by surface morphology control.
Enhancement of adhesive strength of hydroxyapatite films on Ti-29Nb-13Ta-4.6Zr by surface morphology control.
Journal of the mechanical behavior of biomedical materials (2013-01-01)
Junko Hieda, Mitsuo Niinomi, Masaaki Nakai, Ken Cho, Tatsuya Gozawa, Hirokazu Katsui, Rong Tu, Takashi Goto
ABSTRACT
Hydroxyapatite (HAp) films were deposited on a β-type titanium alloy, Ti-29Nb-13Ta-4.6Zr (TNTZ), by metal organic chemical vapor deposition (MOCVD) in order to improve its hard-tissue compatibility. The surface morphologies of TNTZ substrates were changed by acid treatments and mechanical polishing prior to the HAp film deposition. The adhesive strength of the HAp films formed on TNTZ substrates treated with an HF solution increased to twice that of the HAp film deposited on a TNTZ substrate with a mirror-like finish. Complex microstructures with deeply etched grain boundaries, formed on the TNTZ substrates after immersion in the HF solution, were responsible for the increase in the adhesive strength of the HAp film caused by an interlocking effect. The HAp films on TNTZ substrates treated with a H(2)SO(4) solution exhibited lower adhesive strength than HAp films on TNTZ substrates treated with HF solution, regardless of the surface roughness of the substrates. Additionally, acid treatments using HNO(3) and H(2)O(2) solutions did not change the surface morphologies of the TNTZ substrates. The complex microstructures with deeply etched grain boundaries and nanosized asperities formed on the TNTZ substrates are important factors in the improvement of the adhesive strengths of HAp films deposited on TNTZ substrates.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%