MilliporeSigma
  • Study on the waste liquid crystal display treatment: focus on the resource recovery.
Study on the waste liquid crystal display treatment: focus on the resource recovery.
Journal of hazardous materials (2013-01-01)
Xinying Wang, Xuebin Lu, Shuting Zhang
ABSTRACT
A process combined pyrolysis and acid immersion was proposed in this study to dispose the hazardous liquid crystal display (LCD) waste for recovering valuable resources. The thermogravimetric (TG) analysis and fixed bed pyrolysis were investigated for the polarizing film that was separated from LCD. The results suggested the liquid product mainly contained acids, esters and aromatics should be upgraded such as hydrotreating process before used as industrial feedstock or fuel source. The gaseous product mainly consisted of H(2), CO, CO(2) and CH(4) can be used as a valuable fuel. The sulfuric acid immersion experiments were studied for recovering indium from the LCD glass after stripping the polarizing film. Central composite design (CCD) under response surface methodology (RSM) was used to optimize the acid immersion process and the results indicated the indium recovery can be fitted based on the actual value to a polynomial quadratic equation and the temperature was more essential factor than time and acid concentration in the studied ranges. The optimum processing condition was obtained with time 42.2 min, temperature 65.6 °C and acid concentration 0.6 mol/L. Under the optimal conditions, the indium recovery was close to 100%.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)