MilliporeSigma
  • Poly(amido amine)-based mannose-glycodendrimers as multielectron redox probes for improving lectin sensing.
Poly(amido amine)-based mannose-glycodendrimers as multielectron redox probes for improving lectin sensing.
Langmuir : the ACS journal of surfaces and colloids (2013-01-05)
Manuel C Martos-Maldonado, Juan M Casas-Solvas, Indalecio Quesada-Soriano, Luis García-Fuentes, Antonio Vargas-Berenguel
ABSTRACT
An easy-to-prepare series of electroactive poly(amido amine) (PAMAM)-based dendrimers of generations G0 to G2 having mannopyranosylferrocenyl moieties in the periphery to detect carbohydrate-protein interactions is reported. The synthesis involved the functionalization of the PAMAM surface with azidomethylferrocenyl groups and subsequent coupling of mannoside units by the Cu(I)-catalyzed Huisgen reaction. The binding affinity of the series of electroactive glycodendrimers was studied by isothermal titration calorimetry (ITC) and differential pulse voltammetry (DPV). Upon complexation of the glycodendrimers conjugates with prototypical concanavalin A (Con A), voltammograms showed a decrease of the peak current. Such dendrimers showed a notable improvement of redox sensing abilities toward Con A when compared with mono- and divalent analogues, based on both the glycoside multivalent and ferrocene dendritic effects.
Sigma-Aldrich
Ferrocene, 98%
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture