MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Investigation of possible interference by digoxin-like immunoreactive substances on the Architect iDigoxin CMIA in serum samples from pregnant women, and patients with liver disease, renal insufficiency, critical illness, and kidney and liver transplant.
Investigation of possible interference by digoxin-like immunoreactive substances on the Architect iDigoxin CMIA in serum samples from pregnant women, and patients with liver disease, renal insufficiency, critical illness, and kidney and liver transplant.
Clinical laboratory (2013-01-08)
Natalia Lampon, Fernando Pampin, J Carlos Tutor
ABSTRACT
Although the possible interference of digoxin-like immunoreactive substances (DLIS) on the Architect iDigoxin chemiluminiscent microparticle immunoassay (CMIA) has been emphasized by the manufacturer, a specific study about this subject has still not been carried out. Apparent serum digoxin concentrations were determined using the Architect iDigoxin CMIA from Abbott Laboratories in digoxin-free pregnant women (n = 50), and patients with liver disease (n = 50), renal insufficiency (n = 50), kidney (n = 25) or liver (n = 25) transplant, and critical illness (n = 50). In all of the patients included in this study, apparent serum digoxin concentrations were lower than the correspondent quantification limit (< 0.30 microg/L). The Architect iDigoxin CMIA assay would be relatively free from endogenous DLIS positive interferences.
Product Description
Supelco
Digoxin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Digoxin for peak identification, European Pharmacopoeia (EP) Reference Standard
Digoxin, European Pharmacopoeia (EP) Reference Standard
Supelco
Digoxin, analytical standard