MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • New insights into Lactococcus lactis diacetyl- and acetoin-producing strains isolated from diverse origins.
New insights into Lactococcus lactis diacetyl- and acetoin-producing strains isolated from diverse origins.
International journal of food microbiology (2013-01-08)
Delphine Passerini, Valérie Laroute, Michèle Coddeville, Pascal Le Bourgeois, Pascal Loubière, Paul Ritzenthaler, Muriel Cocaign-Bousquet, Marie-Line Daveran-Mingot
ABSTRACT
Lactococcus lactis subsp. lactis biovar diacetylactis strains are used in the dairy industry for generating acetoin and notably diacetyl which imparts a high level of buttery flavor notes. A collection of domesticated and environmental strains was screened for the production of diacetyl or acetoin (D/A), and citrate fermentation. Unexpectedly, both domesticated and environmental strains produced D/A. Domesticated strains belonging to the currently named "biovar diacetylactis" metabolized citrate and produced large amounts of D/A during early growth. They harbored the citP plasmid gene encoding citrate permease and a chromosomal region citM-citI-citCDEFXG involved in citrate metabolism. In these strains, citrate consumption was identified as the major determinant of aroma production. Environmental strains, specifically UCMA5716 and A12, produced as much D/A as the CitP(+) strains, though at slightly lower rates. UCMA5716 was found to contain the citM-citI-citCDEFXG cluster but not the citP gene. A12 had neither. In these strains, production rate of D/A was linearly correlated with pyruvate synthesis rate. However, the correlation factor was strain-dependent, suggesting different modes of regulation for pyruvate rerouting towards fermentation end-products and flavors. This work highlights the genetic and metabolic differences between environmental and domesticated strains. The introduction of environmental strains into industrial processes could considerably increase the diversity of starters, enhancing the delivery of new technological properties.
Product Description
Supelco
2,3-Butanedione, analytical standard
Sigma-Aldrich
2,3-Butanedione, 97%
Sigma-Aldrich
Acetoin, May exist as crystalline dimer
Supelco
Acetoin, analytical standard
Sigma-Aldrich
Acetoin, primarily dimer, ≥95%, FG
Sigma-Aldrich
Acetoin, natural, ≥95%, FG