MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Fluorescent live-cell imaging of metabolically incorporated unnatural cyclopropene-mannosamine derivatives.
Fluorescent live-cell imaging of metabolically incorporated unnatural cyclopropene-mannosamine derivatives.
Chembiochem : a European journal of chemical biology (2013-01-08)
Christian M Cole, Jun Yang, Jolita Šečkutė, Neal K Devaraj
ABSTRACT
Sugar coated: We recently developed methylcyclopropenes as low-molecular-weight tetrazine coupling partners. Here, we demonstrate that methylcyclopropenes can meet the stringent steric demands required for metabolic imaging of unnatural mannosamines on live cells. Using sequential azide-alkyne chemistry, we also demonstrate multicolor imaging of two different metabolically incorporated unnatural sugars.
Sigma-Aldrich
D-Mannosamine hydrochloride, ≥98% (HPLC)