MilliporeSigma
  • STAT6 deficiency ameliorates severity of oxazolone colitis by decreasing expression of claudin-2 and Th2-inducing cytokines.
STAT6 deficiency ameliorates severity of oxazolone colitis by decreasing expression of claudin-2 and Th2-inducing cytokines.
Journal of immunology (Baltimore, Md. : 1950) (2013-01-11)
Michael J Rosen, Rupesh Chaturvedi, M Kay Washington, Lindsay A Kuhnhein, Preston D Moore, Scott S Coggeshall, Elizabeth M McDonough, Jörn-Hendrik Weitkamp, Amar B Singh, Lori A Coburn, Christopher S Williams, Fang Yan, Luc Van Kaer, R Stokes Peebles, Keith T Wilson
ABSTRACT
Patients suffering from ulcerative colitis (UC) exhibit chronic colonic inflammation caused by a dysregulated mucosal immune response and epithelial barrier disruption. Th2 cytokines, including IL-13, have been implicated in the pathogenesis of UC. IL-13 induces phosphorylation of STAT6, and we previously demonstrated increased epithelial p-STAT6 in children with UC. In this study, we investigated the role of STAT6 in oxazolone colitis, a murine model of UC, by inducing colitis in STAT6-deficient (STAT6(-/-)) and wild type (WT) mice. We observed increased epithelial cell, T cell, macrophage, and NKT cell STAT6 phosphorylation, as well as increased p-STAT6(+) IL-13-producing NKT cells, in colitic WT mice. Colitis was attenuated in STAT6(-/-) mice, with improvements in weight, colon length, and histopathology. There was decreased induction of the pore-forming tight junction protein claudin-2 in STAT6(-/-) mice. Similarly, short hairpin RNA STAT6 knockdown reduced claudin-2 induction and transepithelial resistance decrease in IL-13-treated human T84 cells. Tissue expression of IL-13, IFN-γ, IL-17, and IL-10 mRNA was similarly induced in WT and STAT6(-/-) colitic mice; however, we observed increased mRNA expression for the Th2-inducing cytokines IL-33 and thymic stromal lymphopoietin in WT mice with colitis, which was abrogated in STAT6(-/-) mice. Mesenteric lymph node cells from STAT6(-/-) mice with colitis exhibited reduced secretion of IL-4, IL-5, IL-13, and IFN-γ. IL-33 augmented mesenteric lymph node cell secretion of IL-5, IL-13, IL-6, and IFN-γ. These data implicate STAT6 in the pathogenesis of colitis in vivo with important roles in altering epithelial barrier function and regulating Th2-inducing cytokine production.
Sigma-Aldrich
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, purified by recrystallization
Supelco
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, analytical standard, for drug analysis