MilliporeSigma
Curcumin and aging.
BioFactors (Oxford, England) (2013-01-18)
Li-Rong Shen, Laurence D Parnell, Jose M Ordovas, Chao-Qiang Lai
ABSTRACT
Turmeric has been used commonly as a spice, food additive, and an herbal medicine worldwide. Known as a bioactive polyphenolic extract of Turmeric, curcumin has a broad range of health benefit properties for humans. Recently, active research on curcumin with respect to aging and related traits in model organisms has demonstrated that curcumin and its metabolite, tetrahydrocurcumin (THC), increase mean lifespan of at least three model organisms: nematode roundworm, fruit fly Drosophila, and mouse. Nematodes grown on media containing curcumin showed a significantly increased lifespan by reducing the production of reactive oxygen species. Genes osr-1, sek-1, mek-1, skn-1, unc-43, sir-2.1, and age-1 are required for curcumin-mediated lifespan extension. The lifespan extension of Drosophila by curcumin supplementation was associated with increased superoxide dismutase (SOD) activity, and decreased lipofuscin and malondialdehyde levels. Curcumin up-regulated expression of SOD genes and down-regulated expression of several age-related genes, such as dInR, ATTD, Def, CecB, and DptB. In addition, THC extended lifespan in Drosophila and inhibited the oxidative stress response by regulating FOXO and Sir2. Mice fed diets containing THC starting at the age of 13 months had significantly increased mean lifespan. In summary, the positive effects of curcumin on lifespan extension likely arise from beneficial regulation of common oxidative stress responses and age-related genes. Understanding the molecular mechanism(s) of curcumin action has provided base knowledge and rationale for future human clinical trials, and for nutritional intervention in aging and age-associated disorders in humans.
Product Description
Supelco
Tetrahydrocurcumin, analytical standard