MilliporeSigma
  • Chemical and electron microscopic studies of factors associated with the release of penicillinase from Staphylococcus aureus.
Chemical and electron microscopic studies of factors associated with the release of penicillinase from Staphylococcus aureus.
Antonie van Leeuwenhoek (1979-01-01)
T K Kim, J B Hammond, J R Chipley
PMID233290
ABSTRACT
The effect of various factors such as sodium chloride, sodium citrate, pH, linase by Staphylococcus aureus ATCC 14458 was investigated. Penicillinase was measured at selected time intervals from supernates of cultures grown in Antibiotic Medium 3 broth containing various concentrations of salts or buffers or from supernates of cultures treated with lysostaphin and subsequently disrupted by French press treatment. Incubation of cells with media containing either sodium chloride (5, 10, and 15%), sodium citrate (5 and 10%), or organic buffers (Tris-HCl, 2.5, 5.0, and 7.5%; BES, 10 and 20%) resulted in a significant stimulation of the release of penicillinase when compared to control cells. It was also observed that pH 7.0-7.5 was optimal for penicillinase activity and release. From studies of enzymatic and mechanical disruption of cells, it was observed that an increase in ionic strength of the suspending medium to certain optimal levels appeared to stimulate the conversion of penicillinase to an extracellular form. Electron microscopic studies revealed that a large number of mesosomal vesicles seemed to be present in cells incubated for 4 hours in media containing various concentrations of sodium chloride. It is proposed that either appearance of vesicles or convolution of cell membrane, which may be caused by further synthesis of new membrane, is involved in stimulation of the synthesis and release of membrane-bound penicillinase.
Sigma-Aldrich
BES, ≥99.0% (titration)