MilliporeSigma
  • Clinical and research markers of oxidative stress in chronic kidney disease.
Clinical and research markers of oxidative stress in chronic kidney disease.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2013-01-24)
Patrick Steven Tucker, Vincent James Dalbo, Thin Han, Michael Ian Kingsley
ABSTRACT
Kidney-related pathologies have increasing prevalence rates, produce a considerable financial burden, and are characterized by elevated levels of oxidative stress (OS). This review examines relationships between chronic kidney disease (CKD) and markers of OS and antioxidant status (AS). A systematic review of MEDLINE-indexed clinical trials, randomized controlled trials and comparative studies that examined OS and AS was performed. Several markers emerged as well-suited indicators of OS and AS in CKD: malondialdehyde, F2-isoprostanes, lipid hydroperoxides, asymmetric dimethylarginine, 8-oxo-7,8-dihydro-2'-deoxyguanosine, protein carbonyls, advanced oxidation protein products and glutathione-related activity.
Sigma-Aldrich
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)