MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Simultaneous determination of bisphenol A, tetrabromobisphenol A, and perfluorooctanoic acid in small household electronics appliances of "Prohibition on Certain Hazardous Substances in Consumer Products" instruction using ultra-performance liquid chromatography-tandem mass spectrometry with accelerated solvent extraction.
Simultaneous determination of bisphenol A, tetrabromobisphenol A, and perfluorooctanoic acid in small household electronics appliances of "Prohibition on Certain Hazardous Substances in Consumer Products" instruction using ultra-performance liquid chromatography-tandem mass spectrometry with accelerated solvent extraction.
Journal of separation science (2013-01-24)
Qiaozhen Guo, Zhenxia Du, Yun Zhang, Xiaoyu Lu, Jinhua Wang, Wenlian Yu
ABSTRACT
Simultaneous determination of bisphenol A, tetrabromobisphenol A, and perfluorooctanoic acid in small household electronics appliances by accelerated solvent extraction-ultra-performance liquid chromatography-tandem mass spectrometry was established. Samples, heated for 5 min, were extracted by toluene/methanol (10:1, v/v) under the pressure 1500 psi at 100°C, and were extracted 3 static cycles with 20 min per cycle. And then 15 mL extractant solvent was used to wash the samples, and at last the sample was purged by nitrogen for 100 s. The partial extractant (10 mL) was concentrated by nitrogen and re-dissolved with 1 mL methanol/water (1:1, v/v). The three compounds were separated by BEH C18 column effectively in 3 min and detected by electrospray ionization mode mass spectrometry. The linear ranges for bisphenol A, perfluorooctanoic acid, and tetrabromobisphenol A were 1-100, 10-1000 ng/mL, and 0.1-10 μg/mL, respectively. The correlation coefficient was greater than 0.996. The LOD and limit of quantitation for three compounds were 0.1, 10, 1 ng/mL, and 0.5, 50, 5 ng/mL, respectively. And the recoveries were 84-92, 76-82, and 72-74%, respectively, with RSD < 5%. The method was successfully used in determining the real samples. The method and the result were confirmed by liquid chromatography-ion trap-time of flight mass spectrometry.
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Pentadecafluorooctanoic acid ammonium salt, ≥98.0% (sum of isomers)
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
3,3′,5,5′-Tetrabromobisphenol A, 97%