MilliporeSigma
  • Evaluation of different plating media used in the isolation of salmonellas from environmental samples.
Evaluation of different plating media used in the isolation of salmonellas from environmental samples.
The Journal of applied bacteriology (1989-04-01)
M A Moriñigo, E Martinez-Manzanares, A Muñoz, R Cornax, P Romero, J J Borrego
ABSTRACT
Different serotypes of salmonellas were compared for selectivity and efficiency of recovery using 11 plating media. No optimal growth was obtained after 24 h incubation in any of the media, but after 48 h, brilliant green, brilliant green-phenol red-lactose-sucrose, bismuth sulphite, xylose-lysine-deoxycholate and Hektoen enteric agars showed optimal recovery of all the salmonella serotypes. Xylose-lysine-deoxycholate and brilliant green-phenol red-lactose-sucrose agars were the most selective media for all salmonella serotypes. Addition of 10 micrograms/ml of sodium novobiocin to the tryptic soy-xylose-lysine and tryptic soy-brilliant green agars significantly improved their selectivity but reduced or inhibited the growth of some salmonella serotypes, including Salmonella typhi. Xylose-lysine-deoxycholate agar gave the highest recovery percentage of stressed salmonellas with a double-agar layer technique. Good recovery was also obtained on brilliant green-phenol red-lactose-sucrose, tryptic soy-brilliant green, tryptic soy-brilliant green-novobiocin, tryptic, soy-xylose-lysine and tryptic soy-xylose-lysine-novobiocin agars. Salmonella-shigella agar was the least efficient medium for the recovery of salmonellas under stress-induced or non-stressed conditions.