MilliporeSigma
  • Effectiveness of efavirenz-based regimens in young HIV-infected children treated for tuberculosis: a treatment option for resource-limited settings.
Effectiveness of efavirenz-based regimens in young HIV-infected children treated for tuberculosis: a treatment option for resource-limited settings.
PloS one (2013-02-02)
Janneke H van Dijk, Catherine G Sutcliffe, Francis Hamangaba, Christopher Bositis, Douglas C Watson, William J Moss
ABSTRACT
Antiretroviral treatment (ART) options for young children co-infected with HIV and tuberculosis are limited in resource-poor settings due to limited data on the use of efavirenz (EFV). Using available pharmacokinetic data, an EFV dosing schedule was developed for young co-infected children and implemented as the standard of care at Macha Hospital in Southern Province, Zambia. Treatment outcomes in children younger than 3 years of age or weighing less than 10 kg receiving either EFV-based ART plus anti-tuberculous treatment or nevirapine-based (NVP) ART were compared. Treatment outcomes were measured in a cohort of HIV-infected children seeking care at Macha Hospital in rural Zambia from 2007 to 2010. Information on the diagnosis and treatment of tuberculosis was abstracted from medical records. Forty-five children treated for tuberculosis initiated an EFV-based regimen and 69 children initiated a NVP-based regimen, 7 of whom also were treated for tuberculosis. Children receiving both regimens were comparable in age, but children receiving EFV started ART with a lower CD4(+) T-cell percentage and weight-for-age z-score. Children receiving EFV experienced increases in both CD4(+) T-cell percentage and weight-for-age z-score during follow-up, such that levels were comparable to children receiving NVP after two years of ART. Cumulative survival after 12 months of ART did not differ between groups (NVP:87%;EFV:80%;p = 0.25). Eleven children experienced virologic failure during follow-up.The adjusted hazard ratio of virologic failure comparing EFV to NVP was 0.25 (95% CI:0.05,1.24) and 0.13 (95% CI:0.03,0.62) using thresholds of 5000 and 400 copies/mL, respectively.Five children receiving EFV were reported to have had convulsions after ART initiation compared to only one child receiving NVP (p = 0.04). Despite poorer health at ART initiation, children treated for tuberculosis and receiving EFV-based regimens showed significant improvements comparable to children receiving NVP-based regimens. EFV-based regimens should be considered for young HIV-infected children co-infected with tuberculosis in resource-limited settings.
Sigma-Aldrich
Efavirenz, ≥98% (HPLC)