MilliporeSigma
  • Selective bond cleavage in potassium collisions with pyrimidine bases of DNA.
Selective bond cleavage in potassium collisions with pyrimidine bases of DNA.
Physical review letters (2013-02-07)
Diogo Almeida, Filipe Ferreira da Silva, Gustavo García, Paulo Limão-Vieira
ABSTRACT
Electron transfer in alkali-molecule collisions to gas phase thymine and uracil yielding H- formation is selectively controlled in the energy range between 5.3 and 66.1 eV. By tuning the collision energy, electron transfer from the alkali to partly deuterated thymine, methylated thymine at the N1 and methylated uracil at the N3 positions, H- loss proceeds not only through the breaking of the (C-H) against (N-H) bonds but also through N1 against N3 sites. Such selectivity, as far as bond and site are concerned, is here reported for the first time by electron transfer induced dissociation experiments in alkali-molecule collisions.
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymine, ≥99%