MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
[A study on early-onset group "B" streptococcal neonatal infection].
Akusherstvo i ginekologiia (2013-02-09)
R Vacheva, M Todorova, A Decheva, N Yarakova, I Kraleva, Ts Takova, N Dimitrova, A Dobreva
ABSTRACT
The results achieved with 80% reduction in the incidence of early-onset neonatal group B streptococcal (GBS) sepsis following the implementation of the preliminary (1996, 2002) and subsequently the revised (2010) guidelines for intrapartum antibiotic prophylaxis imposed the discussion on a large scale of the updated:--algorithms for GBS screening (35-37 weeks of gestation) with the recommended dosage of penicillin-G for intrapartum antibiotic prophylaxis for women having normal labor and delivery;--algorithms for GBS screening and intrapartum antibiotic prophylaxis for women with preterm labor (PPROM) or premature rupture of membranes (PROM);--intrapartum antibiotic prophylaxis regimens for women with penicillin allergy;--algorithm for management of newborns with respect to risk of early-onset GBS disease. The present study is aimed at studying the distribution of the early-onset GBS disease in our country based on the data of leading obstetrics & gynecology clinics and wards. The aim is to diferrentiate clinically the cases and investigate the influence of the known risk factors on the part of the mother. A special accent is put over the microbiological diagnostics of cases in view of CDC expanded recommendations on the laboratory methods for identification of GBS. As a final conclusion the necessity for introduction of an official registration of the early- and late-onset GBS disease in the country is emphasized.