MilliporeSigma
  • Silicatein conjugation inside nanoconfined geometries through immobilized NTA-Ni(II) chelates.
Silicatein conjugation inside nanoconfined geometries through immobilized NTA-Ni(II) chelates.
Chemical communications (Cambridge, England) (2013-02-09)
Muhammad Nawaz Tahir, Mubarak Ali, Rute Andre, Werner E G Müller, Heinz-Christoph Schröder, Wolfgang Tremel, Wolfgang Ensinger
ABSTRACT
The chemical modification and bioconjugation processes inside confined geometries by His-tagged silicatein promote sensitive changes in the polarity and surface charge density that mainly contribute to the ionic current rectification properties of the single conical nanopores.
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%