MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficacy of Duet dual-action adulticide against caged Aedes albopictus with the use of an ultra-low volume cold aerosol sprayer.
Efficacy of Duet dual-action adulticide against caged Aedes albopictus with the use of an ultra-low volume cold aerosol sprayer.
Journal of the American Mosquito Control Association (2013-02-12)
Devi S Suman, Sean P Healy, Ary Farajollahi, Scott C Crans, Randy Gaugler
ABSTRACT
Duet was field evaluated against caged Aedes albopictus, with the use of a truck-mounted ultra-low volume (ULV) cold aerosol sprayer at the rate of 90.6 ml/ha. Cages were placed in 3 rows stationed 30.5, 61.0, and 91.5 m downwind of the spray vehicle. Initial knockdown was >99% with mortality ranging between 95.8% and 98.0% across the distances. Volume median diameter (VMD) and droplet density were slightly reduced (16.4% and 18.6%, respectively) by distance. Reduction in VMD and droplet density, however, did not reduce mortality. The ULV application of Duet is an effective adulticide against Ae. albopictus and may be a useful tool for use in mosquito abatement programs.
Product Description
Supelco
Piperonylbutoxide, PESTANAL®, analytical standard
Sigma-Aldrich
Piperonylbutoxide, technical grade, 90%
Supelco
Prallethrin, PESTANAL®, analytical standard
Supelco
Phenothrin, PESTANAL®, analytical standard