MilliporeSigma
  • Normal Hb A2 β-thalassemia trait: frameshift mutation (HBB: c.187_251dup) in cis with the Hb A2' δ-globin gene missense mutation (HBD: c.49G>C).
Normal Hb A2 β-thalassemia trait: frameshift mutation (HBB: c.187_251dup) in cis with the Hb A2' δ-globin gene missense mutation (HBD: c.49G>C).
Hemoglobin (2013-02-13)
John S Waye, Barry Eng, Laurie Hellens, Betty-Ann Hohenadel, Lisa M Nakamura, Lynda Walker
ABSTRACT
We report the case of a father and daughter who are heterozygous for a duplication of 65 bp within exon 2 of the β-globin gene, resulting in an altered and truncated β-globin chain that is predicted to be non functional. The β-globin gene mutation is in cis with the common Hb A2 ' missense mutation of the δ-globin gene (HBD: c.49G>C), resulting in β-thalassemia (β-thal) trait with normal levels of Hb A2. This is the second report of this β(0)-thal mutation, and both families were associated with the Hb A2 ' variant and normal levels of Hb A2. Laboratories should be aware of the rare occurrence of β-thal trait with normal levels of Hb A2.
Sigma-Aldrich
Hemoglobin A2, Ferrous Stabilized human, lyophilized powder