MilliporeSigma
  • Polyanionic and polyzwitterionic azobenzene ionic liquid-functionalized silica materials and their chromatographic applications.
Polyanionic and polyzwitterionic azobenzene ionic liquid-functionalized silica materials and their chromatographic applications.
Chemical communications (Cambridge, England) (2013-02-19)
Hongdeng Qiu, Shengxiang Jiang, Makoto Takafuji, Hirotaka Ihara
ABSTRACT
New polyanionic and polyzwitterionic azobenzene ionic liquid-functionalized silica materials were designed based on the preparation of a new polymerizable azobenzene anionic monomer and either its cation-exchange with alkylimidazolium after grafting or the formation of an ionic liquid monomer pair before grafting onto silica.
Sigma-Aldrich
Azobenzene, 98%