MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Transcriptional analysis of the effect of exogenous decanoic acid stress on Streptomyces roseosporus.
Transcriptional analysis of the effect of exogenous decanoic acid stress on Streptomyces roseosporus.
Microbial cell factories (2013-02-26)
Guojian Liao, Qing Liu, Jianping Xie
ABSTRACT
Daptomycin is an important antibiotic against infections caused by drug-resistant pathogens. Its production critically depends on the addition of decanoic acid during fermentation. Unfortunately, decanoic acid (>2.5 mM) is toxic to daptomycin producer, Streptomyces roseosporus. To understand the mechanism underlying decanoic tolerance or toxicity, the responses of S. roseosporus was determined by a combination of phospholipid fatty acid analysis, reactive oxygen species (ROS) measurement and RNA sequencing. Assays using fluorescent dyes indicated a sharp increase in reactive oxygen species during decanoic acid stress; fatty acid analysis revealed a marked increase in the composition of branched-chain fatty acids by approximately 10%, with a corresponding decrease in straight-chain fatty acids; functional analysis indicated decanoic acid stress has components common to other stress response, including perturbation of respiratory functions (nuo and cyd operons), oxidative stress, and heat shock. Interestingly, our transcriptomic analysis revealed that genes coding for components of proteasome and related to treholase synthesis were up-regulated in the decanoic acid -treated cells. These findings represent an important first step in understanding mechanism of decanoic acid toxicity and provide a basis for engineering microbial tolerance.
Sigma-Aldrich
Sodium decanoate, ≥98%
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥98.0%
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG