MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Iodobenzene catalyzed C-H amination of N-substituted amidines using m-chloroperbenzoic acid.
Iodobenzene catalyzed C-H amination of N-substituted amidines using m-chloroperbenzoic acid.
Organic letters (2013-03-01)
Santhosh Kumar Alla, Rapolu Kiran Kumar, Pradeep Sadhu, Tharmalingam Punniyamurthy
ABSTRACT
The oxidative C-H amination of N"-aryl-N'-tosyl/N'-methylsulfonylamidines and N,N'-bis(aryl)amidines has been accomplished using iodobenzene as a catalyst to furnish 1,2-disubstituted benzimidazoles in the presence of mCPBA as a terminal oxidant at room temperature. The reaction is general, and the target products can be obtained in moderate to high yields.
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
Iodobenzene, 98%